Merck
  • One-step synthesis of alpha-chloro acetophenones from acid chlorides and aryl precursors.
One-step synthesis of alpha-chloro acetophenones from acid chlorides and aryl precursors.
Organic letters (2007-01-27)
Jonathan Rosen, Dietrich Steinhuebel, Michael Palucki, Ian Davies
ABSTRACT
A direct and efficient method was developed for the preparation of a variety of substituted acetophenone derivatives from readily available arene precursors and acid chlorides. This method has significant generality and affords access to substitution patterns on aryl rings not directly achievable by Friedel-Crafts chemistry. [reaction: see text].