- Hydrothermal growth of large-scale micropatterned arrays of ultralong ZnO nanowires and nanobelts on zinc substrate.
Hydrothermal growth of large-scale micropatterned arrays of ultralong ZnO nanowires and nanobelts on zinc substrate.
Chemical communications (Cambridge, England) (2006-08-22)
Conghua Lu, Limin Qi, Jinhu Yang, Li Tang, Dayong Zhang, Jiming Ma
PMID16921442
ABSTRACT
Large-scale, ultralong ZnO nanowire and nanobelt arrays with honeycomb-like micropatterns have been fabricated by hydrothermal oxidation of zinc foil in aqueous alkaline (NH4)2S2O8 solutions.
Product Description
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 1m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Zinc, foil, 2m coil, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.9%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.01mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 4mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95+%