Merck
  • Purification and characterization of cassiicolin, the toxin produced by Corynespora cassiicola, causal agent of the leaf fall disease of rubber tree.
Purification and characterization of cassiicolin, the toxin produced by Corynespora cassiicola, causal agent of the leaf fall disease of rubber tree.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-11-23)
Frédéric de Lamotte, Marie-Pierre Duviau, Christine Sanier, Robert Thai, Joël Poncet, Daniel Bieysse, Frédéric Breton, Valérie Pujade-Renaud
ABSTRACT
Cassiicolin, a phytotoxin produced by the necrotrophic fungus Corynespora cassiicola, was purified to homogeneity from a rubber tree isolate. The optimized protocol involves reverse phase chromatography followed by size exclusion chromatography, with monitoring of the toxicity on detached rubber tree leaves. Cassiicolin appeared to be a peptide composed of 27 amino acids, glycosylated on the second residue, with a N-terminal pyroglutamic acid and 6 cysteines involved in disulfide bonds. Its molecular mass was estimated to be 2885 Da. No significant sequence homology with other proteins could be found. The availability of pure toxin in sufficient amount is a prerequisite for its structure determination, which is a key step in the understanding of the aggression mechanism.
Sigma-Aldrich
Pyroglutamate Aminopeptidase from Pyrococcus furiosus, recombinant, expressed in E. coli, ~90% (SDS-PAGE), ≥5.0 units/mg protein
Pricing and availability is not currently available.