Merck
  • Molybdenum oxo-imido aryloxide complexes: oxo analogues of olefin metathesis catalysts.
Molybdenum oxo-imido aryloxide complexes: oxo analogues of olefin metathesis catalysts.
Inorganic chemistry (2006-05-23)
Warren B Cross, James C Anderson, Claire Wilson, Alexander J Blake
ABSTRACT
The novel 16-electron molybdenum oxo-imido bis(aryloxide) complexes [Mo(NtBu)(O)(2,6-Me2C6H3O)2(py)] (1) and [Mo(NtBu)(O)(2,6-iPr2C6H3O)2(py)] (2) have been prepared by the salt elimination reactions of [Mo(NtBu)(O)Cl2(DME)] with the appropriate lithium aryloxide and from the cycloaddition reactions of tert-butyl isocyanate with the appropriate molybdenum dioxo bis(aryloxide) complex [Mo(O)2(OAr)2(py)n]. Complexes 1 and 2 are the first isolable and crystallographically characterized molybdenum oxo-imido aryloxide complexes. The geometry around the metal in complexes 1 and 2 is best described as a distorted trigonal bipyramid, with the imido and pyridine ligands occupying the axial positions and the oxo and aryloxide ligands in the equatorial plane. X-ray and IR data have confirmed that the imido ligand is the dominant pi donor in the complexes, resulting in an Mo-O bond order of less than 2.5. Reaction of [Mo(NtBu)(O)Cl2(DME)] with Li(OCH2tBu) instead gave the novel complex [Mo(NtBu)(OCH2tBu)3Cl(py)] (3).
Sigma-Aldrich
tert-Butyl isocyanate, 97%