Merck
  • Synthesis and biological activity of some derivatives of thiochroman-4-one and tetrahydrothiapyran-4-one.
Synthesis and biological activity of some derivatives of thiochroman-4-one and tetrahydrothiapyran-4-one.
Journal of medicinal chemistry (1977-06-01)
K Ramalingam, G X Thyvelikakath, K D Berlin, R W Chesnut, R A Brown, N N Durham, S E Ealick, D van der Helm
PMID406397
ABSTRACT
A small series of pyrazoles and isoxazoles derived from thiochroman-4-one has been synthesized and characterized. The compounds were examined for their in vitro inhibitory activity against Bacillus subtilis and Pseudomonas fluorescens. Among the tested compounds the pyrazole derivative from thiochroman-4-one was found to be the most effective inhibitor of growth of B. subtilis. Extensive H NMR analysis was recorded for all compounds.
Sigma-Aldrich
Thiochroman-4-one, 97%