Merck
  • Pan-PPAR agonist beneficial effects in overweight mice fed a high-fat high-sucrose diet.
Pan-PPAR agonist beneficial effects in overweight mice fed a high-fat high-sucrose diet.
Nutrition (Burbank, Los Angeles County, Calif.) (2009-03-10)
Caroline Fernandes-Santos, Rafael Evangelista Carneiro, Leonardo de Souza Mendonca, Marcia Barbosa Aguila, Carlos Alberto Mandarim-de-Lacerda
ABSTRACT
We analyzed the effect of peroxisome proliferator-activated receptor (PPAR) agonists on adipose tissue morphology, adiponectin expression, and its relation to glucose and insulin levels in C57BL/6 mice fed a high-fat high-sucrose (HFHS) diet. Male C57BL/6 mice received one of five diets: standard chow, HFHS chow, or HFHS plus rosiglitazone (PPAR-gamma agonist), fenofibrate (PPAR-alpha agonist), or bezafibrate (pan-PPAR agonist). Diets were administered for 11 wk and medications from week 6 to week 11. Glucose intolerance (GI) and insulin resistance were evaluated by oral glucose tolerance testing and homeostasis model assessment for insulin resistance, respectively. Adipocyte diameter was analyzed in epididymal, inguinal, and retroperitoneal fat pads and by adiponectin immunostain. Mice fed the HFHS chow had hyperglycemia, GI, insulin resistance, increased fat pad weight, adipocyte hypertrophy, and decreased adiponectin immunostaining. Rosiglitazone improved GI, insulin sensitiveness, and adiponectin immunostaining, but it resulted in body weight gain, hyperphagia, and adipocyte and heart hypertrophy. Fenofibrate improved all parameters except for fasting glucose and GI. Bezafibrate was the most efficient in decreasing body weight and glucose intolerance. Activation of PPAR-alpha, -delta, and -gamma together is better than the activation of PPAR-alpha or -gamma alone, because bezafibrate showed a wider range of action on metabolic, morphologic, and biometric alterations due to an HFHS diet in mice.