Merck
  • Determination of α-keto acids in pork meat and Iberian ham via tandem mass spectrometry.
Determination of α-keto acids in pork meat and Iberian ham via tandem mass spectrometry.
Food chemistry (2013-04-13)
Francisco J Hidalgo, José L Navarro, Rosa M Delgado, Rosario Zamora
ABSTRACT
An analytical method which offers accurate determination and identification of eight α-keto acids (α-ketoglutaric acid, pyruvic acid, 4-hydroxyphenylpyruvic acid, 3-methyl-2-oxobutyric acid, α-keto-γ-methylthiobutyric acid, 4-methyl-2-oxovaleric acid, 3-methyl-2-oxovaleric acid, and phenylpyruvic acid) in pork meat and Iberian ham samples is reported. The method utilises a highly selective and sensitive method of multiple reaction monitoring (MRM) by mass spectrometry. The analytical method is simple (although the chemical derivatisation of the α-keto acids with dansylhydrazine is required), precise (<18% RSD), accurate (90-110%), sensitive (0.01-0.34 mg/kg of defatted and freeze-dried meat depending on the α-keto acid) and linear (R>0.99) over several orders of magnitude (until 0.01-146.1 mg/kg of defatted and freeze-dried meat depending on the α-keto acid). Using this methodology, α-keto acids were found to be present in pork meat to a low extent, and their concentration increased when they were determined in Iberian ham. This is the first report of the presence of α-keto acids in both pork meats and Iberian hams.
Sigma-Aldrich
4-Hydroxyphenylpyruvic acid, 98%
Pricing and availability is not currently available.