Merck
  • Influence of water storage and thermal cycling on the fracture strength of all-porcelain, resin-bonded fixed partial dentures.
Influence of water storage and thermal cycling on the fracture strength of all-porcelain, resin-bonded fixed partial dentures.
The Journal of prosthetic dentistry (1994-03-01)
M Kern, T Fechtig, J R Strub
ABSTRACT
The purpose of this in vitro study was to evaluate the influence of water storage and thermal cycling on the fracture strength of two designs of all-porcelain, resin-bonded fixed partial dentures made with the glass-infiltrated alumina ceramic In-Ceram. Forty frameworks of In-Ceram ceramic veneered with Vitadur-N porcelain were made to replace an upper central incisor bonded to extracted natural teeth. In 20 of the restorations the pontic was veneered only labially with Vitadur-N porcelain and no proximal box preparations were made (design A). The other 20 restorations had additional box preparations proximal to the pontic and the In-Ceram ceramic pontic framework was shifted to the labial aspect and veneered circumferentially with Vitadur-N porcelain (design B). Subgroups of 10 restorations were stored for 7 days in thymol solution or alternatively for 150 days in artificial saliva with thermal cycling for 18,750 cycles between 5 degrees and 55 degrees C. Restorations with design B had a statistically significantly higher fracture strength than restorations with design A. Storage in artificial saliva with thermal cycling significantly reduced the fracture strength of the resin-bonded In-Ceram fixed partial dentures. Therefore, the long-term clinical behavior of resin-bonded In-Ceram fixed partial dentures must be evaluated before general clinical use can be recommended.
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Thymol, analytical standard
Sigma-Aldrich
Thymol, ≥98.5%
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Thymol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%