Merck
  • Both high and low levels of blood vitamin D are associated with a higher prostate cancer risk: a longitudinal, nested case-control study in the Nordic countries.
Both high and low levels of blood vitamin D are associated with a higher prostate cancer risk: a longitudinal, nested case-control study in the Nordic countries.
International journal of cancer (2003-11-18)
Pentti Tuohimaa, Leena Tenkanen, Merja Ahonen, Sonja Lumme, Egil Jellum, Göran Hallmans, Pär Stattin, Sverre Harvei, Timo Hakulinen, Tapio Luostarinen, Joakim Dillner, Matti Lehtinen, Matti Hakama
ABSTRACT
Vitamin D inhibits the development and growth of prostate cancer cells. Epidemiologic results on serum vitamin D levels and prostate cancer risk have, however, been inconsistent. We conducted a longitudinal nested case-control study on Nordic men (Norway, Finland and Sweden) using serum banks of 200,000 samples. We studied serum 25(OH)-vitamin D levels of 622 prostate cancer cases and 1,451 matched controls and found that both low (</=19 nmol/l) and high (>/=80 nmol/l) 25(OH)-vitamin D serum concentrations are associated with higher prostate cancer risk. The normal average serum concentration of 25(OH)-vitamin D (40-60 nmol/l) comprises the lowest risk of prostate cancer. The U-shaped risk of prostate cancer might be due to similar 1,25-dihydroxyvitamin D(3) availability within the prostate: low vitamin D serum concentration apparently leads to a low tissue concentration and to weakened mitotic control of target cells, whereas a high vitamin D level might lead to vitamin D resistance through increased inactivation by enhanced expression of 24-hydroxylase. It is recommended that vitamin D deficiency be supplemented, but too high vitamin D serum level might also enhance cancer development.
Product Description
Ergocalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergocalciferol, 40,000,000 USP units/g
Sigma-Aldrich
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
Supelco
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material