Merck
Oxidative stress in patients with multiple sclerosis.
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (1999-12-28)
C Syburra, S Passi
ABSTRACT
It is well known that brain and nervous system cells are prone to oxidative damage because of their relatively low content of antioxidants, especially enzymatic ones, and of the high levels of both membrane polyunsaturated fatty acids (PUFA) and iron easily released from injured cells. We have investigated the oxidative stress in the blood (plasma, erythrocytes and lymphocytes) of 28 patients affected with multiple sclerosis (MS) and of 30 healthy age matched controls, by performing a multiparameter analysis of non-enzymatic and enzymatic antioxidants--Vitamin E (Vit. E), ubiquinone (UBI), reduced and oxidized glutathione (GSH, GS-SG), superoxide dismutase (SOD), glutathione peroxidase (GPX), catalase (CAT) and fatty acid patterns of phospholipids (PL-FA). PL-FA and Vit. E were assayed by GC-MS; UBI and GSH/GS-SG by HPLC; SOD, GPX and CAT by spectrophotometry. In comparison to controls, patients with MS showed significantly reduced levels of plasma UBI (0.21 +/- 0.10 vs. 0.78 +/- 0.08 mg/ml, p < 0.001), plasma Vit. E (7.4 +/- 2.1 vs. 11.4 +/- 1.8 mg/ml, p < 0.01), lymphocyte UBI (8.1 +/- 4.0 vs. 30.3 +/- 7.2 ng/ml blood, p < 0.001) and erythrocyte GPX (22.6 +/- 5.7 vs. 36.3 +/- 6.4 U/g Hb, p < 0.001). This blood antioxidant deficiency was associated with plasma levels of PL-PUFA--especially C20:3 n-6 and C20:4 n-6--significantly higher than controls. In conclusion, the blood of patients with MS shows the signs of a significant oxidative stress. The possibility of counteracting it by antioxidant administration plus an appropriate diet, might represent a promising way of inhibiting the progression of the disease. Antioxidant supplements should include not only GSH repleting agents, but also Vit. E, ubiquinol, and selenium.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
Acetic acid, analytical standard
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Tin, ≥99%, powder