Merck
  • NOTCH1 and NOTCH3 coordinate esophageal squamous differentiation through a CSL-dependent transcriptional network.
NOTCH1 and NOTCH3 coordinate esophageal squamous differentiation through a CSL-dependent transcriptional network.
Gastroenterology (2010-08-31)
Shinya Ohashi, Mitsuteru Natsuizaka, Yumi Yashiro-Ohtani, Ross A Kalman, Momo Nakagawa, Lizi Wu, Andres J Klein-Szanto, Meenhard Herlyn, J Alan Diehl, Jonathan P Katz, Warren S Pear, John T Seykora, Hiroshi Nakagawa
ABSTRACT
The Notch receptor family regulates cell fate through cell-cell communication. CSL (CBF-1/RBP-jκ, Su(H), Lag-1) drives canonical Notch-mediated gene transcription during cell lineage specification, differentiation, and proliferation in the hematopoietic system, the intestine, the pancreas, and the skin. However, the functional roles of Notch in esophageal squamous epithelial biology are unknown. Normal esophageal keratinocytes were stimulated with calcium chloride to induce terminal differentiation. The squamous epithelia were reconstituted in organotypic 3-dimensional culture, a form of human tissue engineering. Notch was inhibited in culture with a γ-secretase inhibitor or dominant negative mastermind-like 1 (DNMAML1). The roles of Notch receptors were evaluated by in vitro gain-of-function and loss-of-function experiments. Additionally, DNMAML1 was targeted to the mouse esophagus by cytokeratin K14 promoter-driven Cre (K14Cre) recombination of Lox-STOP-Lox-DNMAML1. Notch-regulated gene expression was determined by reporter transfection, chromatin immunoprecipitation assays, quantitative reverse-transcription polymerase chain reaction, Western blotting, immunofluorescence, and immunohistochemistry. NOTCH1 (N1) was activated at the onset of squamous differentiation in the esophagus. Intracellular domain of N1 (ICN1) directly activated NOTCH3 (N3) transcription, inducing HES5 and early differentiation markers such as involucrin (IVL) and cytokeratin CK13 in a CSL-dependent fashion. N3 enhanced ICN1 activity and was required for squamous differentiation. Loss of Notch signaling in K14Cre;DNMAML1 mice perturbed esophageal squamous differentiation and resulted in N3 loss and basal cell hyperplasia. Notch signaling is important for esophageal epithelial homeostasis. In particular, the cross talk of N3 with N1 during differentiation provides novel, mechanistic insights into Notch signaling and squamous epithelial biology.
Sigma-Aldrich
Anti-RBP-Jk Antibody, Chemicon®, from rabbit
Sigma-Aldrich
γ-Secretase Inhibitor XXI, Compound E, This g-secretase inhibitor, CAS 209986-17-4, is a cell-permeable, potent, selective, non-transition-state analog inhibitor of γ-secretase and Notch processing. Lowers Aβ levels in APP transgenic mice