Merck
  • Evidence for a role of collagen synthesis in arterial smooth muscle cell migration.
Evidence for a role of collagen synthesis in arterial smooth muscle cell migration.
The Journal of clinical investigation (1998-06-13)
E F Rocnik, B M Chan, J G Pickering
ABSTRACT
Migration of smooth muscle cells (SMCs) and collagen synthesis by SMCs are central to the pathophysiology of vascular disease. Both processes can be induced shortly after vascular injury; however, a functional relationship between them has not been established. In this study, we determined if collagen synthesis was required for SMC migration, using ethyl-3,4-dihydroxybenzoate (EDHB), an inhibitor of prolyl-4-hydroxylase, and 3,4-DL-dehydroproline (DHP), a proline analogue, which we demonstrate inhibit collagen elaboration by porcine arterial SMCs. SMCs exposed to EDHB or DHP attached normally to collagen- and vitronectin-coated substrates; however, spreading on collagen but not vitronectin was inhibited. SMC migration speed, quantified by digital time-lapse video microscopy, was significantly and reversibly reduced by EDHB and DHP. Flow cytometry revealed that expression of beta1 integrins, through which SMCs interact with collagen, was unaffected by EDHB or DHP. However, both inhibitors prevented normal clustering of beta1 integrins on the surface of SMCs, consistent with a lack of appropriate matrix ligands for integrin engagement. Moreover, there was impaired recruitment of vinculin into focal adhesion complexes of spreading SMCs and disassembly of the smooth muscle alpha-actin-containing cytoskeleton. These findings suggest that de novo collagen synthesis plays a role in SMC migration and implicates a mechanism whereby newly synthesized collagen may be necessary to maintain the transcellular traction system required for effective locomotion.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
3,4-Dehydro-DL-proline, 98%
Sigma-Aldrich
Protocatechuic acid ethyl ester, 97%