Merck
Nitrous oxide and oxygen sedation: an update.
Dental assistant (Chicago, Ill. : 1994) (2013-09-06)
Ann Brunick, Morris S Clark
ABSTRACT
This course will teach the desirable characteristics of nitrous oxide, indications and contraindications for N2O/O2 use, as well as facts and myths surrounding chronic exposure to nitrous oxide, the biologic effects associated with high levels of the gas, and ways to assess and minimize trace gas contamination in an outpatient setting.
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material