Merck
  • In vitro integration of ribosomal RNA synthesis, ribosome assembly, and translation.
In vitro integration of ribosomal RNA synthesis, ribosome assembly, and translation.
Molecular systems biology (2013-06-27)
Michael C Jewett, Brian R Fritz, Laura E Timmerman, George M Church
ABSTRACT
Purely in vitro ribosome synthesis could provide a critical step towards unraveling the systems biology of ribosome biogenesis, constructing minimal cells from defined components, and engineering ribosomes with new functions. Here, as an initial step towards this goal, we report a method for constructing Escherichia coli ribosomes in crude S150 E. coli extracts. While conventional methods for E. coli ribosome reconstitution are non-physiological, our approach attempts to mimic chemical conditions in the cytoplasm, thus permitting several biological processes to occur simultaneously. Specifically, our integrated synthesis, assembly, and translation (iSAT) technology enables one-step co-activation of rRNA transcription, assembly of transcribed rRNA with native ribosomal proteins into functional ribosomes, and synthesis of active protein by these ribosomes in the same compartment. We show that iSAT makes possible the in vitro construction of modified ribosomes by introducing a 23S rRNA mutation that mediates resistance against clindamycin. We anticipate that iSAT will aid studies of ribosome assembly and open new avenues for making ribosomes with altered properties.
Product Description
Clindamycin hydrochloride, European Pharmacopoeia (EP) Reference Standard