Merck
  • Conversion from twice-daily tacrolimus capsules to once-daily extended-release tacrolimus (LCPT): a phase 2 trial of stable renal transplant recipients.
Conversion from twice-daily tacrolimus capsules to once-daily extended-release tacrolimus (LCPT): a phase 2 trial of stable renal transplant recipients.
Transplantation (2013-05-30)
A Osama Gaber, Rita R Alloway, Kenneth Bodziak, Bruce Kaplan, Suphamai Bunnapradist
ABSTRACT
LCP-Tacro is an extended-release formulation of tacrolimus designed for once-daily dosing. Phase 1 studies demonstrated greater bioavailability to twice-daily tacrolimus capsules and no new safety concerns. In this phase 2 study, adult stable kidney transplant patients on tacrolimus capsules (Prograf) twice-daily were converted to tacrolimus tablets (LCP-Tacro) once-daily; patients continued on LCP-Tacro once-daily for days 8 to 21; trough levels were to be maintained between 5 and 15 ng/mL; 24-hr pharmacokinetic assessments were done on days 7 (baseline pre-switch), 14, and 21. Forty-seven patients completed LCP-Tacro dosing per protocol. The mean conversion ratio was 0.71. Pharmacokinetic data demonstrated consistent exposure (AUC) at the lower conversion dose. C(max) (P = 0.0001), C(max)/C(min) ratio (P < 0.001), percent fluctuation (P < 0.0001), and swing (P = 0.0004) were significantly lower and T(max) significantly (P < 0.001) longer for LCP-Tacro versus Prograf. AUC24 and C(min) correlation coefficients after 7 and 14 days of therapy were 0.86 or more, demonstrating a robust correlation between LCP-Tacro tacrolimus exposure and trough levels. There were three serious adverse events; none were related to study drug and all were resolved. Stable kidney transplant patients can be safely converted from Prograf twice-daily to LCP-Tacro. The greater bioavailability of LCP-Tacro allows for once-daily dosing and similar (AUC) exposure at a dose approximately 30% less than the total daily dose of Prograf. LCP-Tacro displays flatter kinetics characterized by significantly lower peak-trough fluctuations.
Product Description
Supelco
Tacrolimus solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®