Merck
  • Use of levonorgestrel intrauterine system for medical indications in adolescents.
Use of levonorgestrel intrauterine system for medical indications in adolescents.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine (2013-04-03)
Lisa L Bayer, Paula J Adams Hillard
ABSTRACT
The levonorgestrel intrauterine system (LNG-IUS) is an underused contraceptive method in adolescent populations. In addition to being a highly effective, reversible, long-acting contraception, the LNG-IUS has many noncontraceptive health benefits including reduced menstrual bleeding, decreased dysmenorrhea and pelvic pain related to endometriosis, and menstruation suppression in teens with physical or developmental disabilities. The LNG-IUS can also provide endometrial protection in teens with chronic anovulation, and may be used to treat endometrial hyperplasia and cancer. This review examines the evidence supporting the use of the LNG-IUS in adolescents for these noncontraceptive benefits.
Product Description
Levonorgestrel, European Pharmacopoeia (EP) Reference Standard
USP
Levonorgestrel, United States Pharmacopeia (USP) Reference Standard
Levonorgestrel for system suitability 2, European Pharmacopoeia (EP) Reference Standard
Supelco
D(−)-Norgestrel, analytical standard