Merck
[Chemical constituents from Hypericum perforatum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-12-14)
Jie Ma, Jianbo Yang, Tengfei Ji, Aiguo Wang, Yalun Su
ABSTRACT
To study the chemical constituents from Hypericum perforatum. Compouds were isolated by chromatographic techniques. Their structures were identified by spectral methods. The inhibitory activity of recombinant human PTP1B was evaluated. Nine compounds were elucidated as D-Mannitol (1), 1,2-benzenedicarboxylic acid bis(1-methylpropyl) ester (2), (7E, 6R,9S)-9-hydroxy-4,7-megastigmadien-3-one (3), (6S,9R)-roseoside (4) , 2,6-dimethoxy-4-hydroquinone-l-O-beta-D-glucopyranoside (5), 2,6-di-methoxy-4-hydroxybenzyl alcohol 1 -O-beta-D-glucopyranoside (6), syringate 4-O-beta-glucopyranoside (7), hypericin (8), skyrin-6-0-beta-D-glucopyranoside (9) , (R)-2,3-dihydroxypropyl 3,4-dihydroxy-benzoate (10). At a concentration of 2 micromol x L(-1), compound 8 inhibited recombinant human PTP1B with inhibitory rate of 96.4% and IC50 of 2.5 micromol x L(-1). compound 10 was new, compounds 2-4, 7 were obtained from Hypericum for the first time and compounds 5-6 was isolated from this plant for the first time. Compound 8 showed remarkable PTP1B inhibitory activity.
Sigma-Aldrich
Skyrin, ≥98% (HPLC), from Penicillium islandicum