Merck
  • Comparing marginal structural models to standard methods for estimating treatment effects of antihypertensive combination therapy.
Comparing marginal structural models to standard methods for estimating treatment effects of antihypertensive combination therapy.
BMC medical research methodology (2012-08-08)
Tobias Gerhard, Joseph Ac Delaney, Rhonda M Cooper-Dehoff, Jonathan Shuster, Babette A Brumback, Julie A Johnson, Carl J Pepine, Almut G Winterstein
ABSTRACT
Due to time-dependent confounding by blood pressure and differential loss to follow-up, it is difficult to estimate the effectiveness of aggressive versus conventional antihypertensive combination therapies in non-randomized comparisons. We utilized data from 22,576 hypertensive coronary artery disease patients, prospectively enrolled in the INternational VErapamil-Trandolapril STudy (INVEST). Our post-hoc analyses did not consider the randomized treatment strategies, but instead defined exposure time-dependently as aggressive treatment (≥3 concomitantly used antihypertensive medications) versus conventional treatment (≤2 concomitantly used antihypertensive medications). Study outcome was defined as time to first serious cardiovascular event (non-fatal myocardial infarction, non-fatal stroke, or all-cause death). We compared hazard ratio (HR) estimates for aggressive vs. conventional treatment from a Marginal Structural Cox Model (MSCM) to estimates from a standard Cox model. Both models included exposure to antihypertensive treatment at each follow-up visit, demographics, and baseline cardiovascular risk factors, including blood pressure. The MSCM further adjusted for systolic blood pressure at each follow-up visit, through inverse probability of treatment weights. 2,269 (10.1%) patients experienced a cardiovascular event over a total follow-up of 60,939 person-years. The HR for aggressive treatment estimated by the standard Cox model was 0.96 (95% confidence interval 0.87-1.07). The equivalent MSCM, which was able to account for changes in systolic blood pressure during follow-up, estimated a HR of 0.81 (95% CI 0.71-0.92). Using a MSCM, aggressive treatment was associated with a lower risk for serious cardiovascular outcomes compared to conventional treatment. In contrast, a standard Cox model estimated similar risks for aggressive and conventional treatments. Clinicaltrials.gov Identifier: NCT00133692.
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder