Merck
  • Evaluating the relationship of methadone concentrations and EDDP formation in chronic pain patients.
Evaluating the relationship of methadone concentrations and EDDP formation in chronic pain patients.
Journal of analytical toxicology (2012-04-19)
Edijs Leimanis, Brookie M Best, Rabia S Atayee, Amadeo J Pesce
ABSTRACT
Methadone is used to treat moderate to severe pain in patients not responsive to non-narcotic analgesics and for maintenance treatment of opioid addiction. Methadone is primarily metabolized by N-demethylation to an inactive metabolite 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidene (EDDP) by CYP3A4 and CYP2B6. Establishing expected concentrations for metabolism of methadone to EDDP using urine excretion data may be useful for monitoring "medications" and toxicity. Urine specimens from chronic pain patients were collected during routine clinic visits. Methadone and EDDP were quantified by liquid chromatography-tandem mass spectrometry. Approximately 8,000 subjects who reported taking methadone had creatinine concentrations ≥20 mg/dL, and excreted concentrations of methadone and EDDP above ≥100 ng/mL were selected. The median methadone urine concentration was 3.03 mg/g cr. Ninety-five percent of the population had concentrations between 0.175 and 20.9 mg/g cr. EDDP was, on average, twice the methadone concentration. The wide variance in relationship of methadone to its metabolite was not concentration-dependent. Variability between subjects was larger than variability within subjects. As the urinary pH increased, the proportion of excreted EDDP increased, implying a preferred excretion of EDDP.
Product Description
Supelco
EDDP perchlorate, ampule of 10 mg, certified reference material, Cerilliant®
Supelco
EDDP perchlorate solution, 1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®