Merck
  • Ciprofloxacin photosensitized oxidation of 2'-deoxyguanosine-5'-monophosphate in neutral aqueous solution.
Ciprofloxacin photosensitized oxidation of 2'-deoxyguanosine-5'-monophosphate in neutral aqueous solution.
Photochemistry and photobiology (2012-02-11)
Yancheng Liu, Peng Zhang, Haixia Li, Wenfeng Wang
ABSTRACT
Laser flash photolysis studies have been carried out to investigate the reactions of ciprofloxacin (CPX) with 2'-deoxyguanosine-5'-monophosphate (dGMP), N, N, N', N'-tetramethyl-p-phenylenediamine (TMPD) and ferulic acid (FCA) in neutral aqueous solutions, respectively. CPX triplet state ((3)CPX*) can be quenched by TMPD, FCA and dGMP, with rate constants of 1.8 × 10(9), 1.5 × 10(9) and 5.8 × 10(7) dm(3) mol(-1) s(-1), respectively. TMPD radical cation (TMPD(·+)) and FCA radical cation (FCA(·+)) were observed directly. The formation rate of CPX radical anion (CPX(·-)) was determined to be 1.5 × 10(9) dm(3) mol(-1) s(-1). Redox reaction of dGMP was investigated through competing reactions using TMPD and FCA as probe. The triplet energy of CPX was determined to be 262 kJ mol(-1). Electron transfer from TMPD, FCA and dGMP to (3)CPX* was proposed.