Merck
  • Fluorine probes for investigating the mechanism of activation of indeno[1,2,3-cd]pyrene to a tumorigenic agent.
Fluorine probes for investigating the mechanism of activation of indeno[1,2,3-cd]pyrene to a tumorigenic agent.
Carcinogenesis (1990-11-01)
J E Rice, E H Weyand, C Burrill, E J LaVoie
ABSTRACT
Indeno[1,2,3-cd]pyrene (IP) is a non-alternant polycyclic aromatic hydrocarbon that has tumor-initiating activity on mouse skin and is carcinogenic in newborn mice and in rat lungs. Previous studies have shown that 8- and 9-hydroxyIP and IP-1,2-diol are major metabolites formed in vivo in mouse skin. 8-HydroxyIP-1,2-diol and 9-hydroxyIP-1,2-diol are also observed as in vivo metabolites of IP. Although 8-hydroxyIP had marginal tumor-initiating activity on mouse skin, IP-1,2-diol and its epoxide precursor, IP-1,2-oxide, had similar tumorigenic activity as IP. In the present study fluorine probes have been employed to investigate the contribution of metabolic activation at the 1,2 and 7-10 positions of IP. At a total initiating dose of 4.0 mumol, 2-fluoroIP induced skin tumors in 76% of the treated animals with an average of 3.9 tumors/mouse. At the same dose, IP induced a 72% incidence of tumor-bearing mice with 2.1 tumors/mouse. In contrast, 8,9-difluoroIP elicited a tumorigenic response in 40% of the treated animals with 0.6 tumors/animal. Five mice from each experimental group were killed at the conclusion of the initiation phase of the bioassay and DNA was isolated from the treated areas of skin. 32P-Postlabeling analysis of the hydrolyzed DNA indicated that IP forms one major detectable DNA adduct that migrates close to the origin. This adduct is absent in mice treated with 8,9-difluoroIP. In contrast, 2-fluoroIP forms one major adduct spot with different retention behavior as compared with the adduct formed from IP. DNA from mice treated topically with IP-1,2-diol and IP-1,2-oxide was subjected to 32P-postlabeling analysis. IP-1,2-diol forms one major DNA adduct spot with mobility similar to that observed for the IP-DNA adduct. IP-1,2-oxide displayed an intense pattern of DNA adducts centered around the location of the IP-DNA adduct. No adducts were detected which had mobility similar to that formed from 2-fluoroIP. These results are consistent with IP undergoing metabolic activation at positions 7-10 either alone or in conjunction with dihydrodiol formation at the 1,2 position.
Product Description
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard