Merck
  • Protective effects of ginsenoside Rd against okadaic acid-induced neurotoxicity in vivo and in vitro.
Protective effects of ginsenoside Rd against okadaic acid-induced neurotoxicity in vivo and in vitro.
Journal of ethnopharmacology (2011-09-29)
Ling Li, Juanfang Liu, Xiaodong Yan, Kefeng Qin, Ming Shi, Tian Lin, Yi Zhu, Tao Kang, Gang Zhao
ABSTRACT
Panax ginseng, a traditional Chinese herbal medicine, has been widely used to restore the disease and enhance the healthy body in Asia for about 5000 years. The present study aimed to investigate the possible neuroprotective effects of ginsenoside Rd against OA-induced toxicity. Ginsenoside Rd was used in tauopahy models of Alzheimer's disease (AD). To mimic the in vivo or in vitro tau hyperphosphorylation, okadaic acid (OA), a protein phosphatase inhibitor, was bilaterally micro-infused into the cerebral ventricle of adult male Sprague-Dawley (SD) rats, or added in media of cultured cortical neurons. The phosphorylation levels of tau and the activities of protein phosphatase 2A (PP-2A) were measured and compared with ginsenoside Rd pretreated groups. Pretreatment with ginsenoside Rd in SD rats (10mg/kg for 7 days) or in cultured cortical neurons (2.5 or 5μmol/L for 12h) reduced OA-induced neurotoxicity and tau hyperphosphorylation by enhancing the activities of PP-2A. The result of the present work implied that ginsenoside Rd protected SD rats and cultured cortical neurons against OA-induced toxicity. The possible neuroprotective mechanism may be that ginsenoside Rd decreases OA-induced the hyperphosphorylation of tau by the increase in activities of PP-2A. Thus, this study promises that ginsenoside Rd might be a potential preventive drug candidate for AD and other tau pathology-related neuronal degenerative diseases.
Product Description
Supelco
Ginsenoside Rd, analytical standard