Merck
  • Staphylococcal enterotoxin B causes proliferation of sensory C-fibers and subsequent enhancement of neurogenic inflammation in rat skin.
Staphylococcal enterotoxin B causes proliferation of sensory C-fibers and subsequent enhancement of neurogenic inflammation in rat skin.
The Journal of infectious diseases (2011-01-22)
Mihoko Ohshima, Mio Miyake, Masanori Takeda, Michihiro Kamijima, Tatsuo Sakamoto
ABSTRACT
Staphylococcal enterotoxin B (SEB) may be associated with the exacerbation of atopic dermatitis. We investigated whether SEB causes proliferation of sensory C-fibers and subsequent enhancement of plasma leakage induced by sensorineural stimulation in rat skin. SEB was applied intracutaneously to the abdomen of preweaning and adult rats. Evans blue dye leakage into the skin induced by topical 10% formalin was measured as an index of neurogenic skin vascular permeability. Local expression of substance P, tachykinin NK1 receptors, and nerve growth factor was assessed immunohistochemically. In addition, we assessed the effects of topical tacrolimus on these skin responses induced by SEB. Increased neurogenic skin plasma leakage was seen 7 days after SEB treatment in 2 different age groups. Innervation of substance P-immunoreactive nerves and expression of tachykinin NK1 receptors and nerve growth factor were also promoted by SEB, peaking at 7 days, 7 days, and 56 h after SEB treatment, respectively. Tacrolimus markedly inhibited these skin changes. SEB increased the innervation of sensory C-fibers and tachykinin NK1 receptors in rat skin, probably because of upregulated production of neurotrophins, including nerve growth factor, leading to enhancement of neurogenic skin inflammation. T cell activation induced by SEB may initiate these changes.
Sigma-Aldrich
Fluocinonide, ≥98% (HPLC)