Merck
  • Development and validation of a LC-MS/MS method for the determination of clebopride and its application to a pharmacokinetics study in healthy Chinese volunteers.
Development and validation of a LC-MS/MS method for the determination of clebopride and its application to a pharmacokinetics study in healthy Chinese volunteers.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-07-06)
Zhirong Tan, Dongsheng Ouyang, Yao Chen, Gan Zhou, Shan Cao, Yicheng Wang, Xiujuan Peng, Honghao Zhou
ABSTRACT
A sensitive and specific liquid chromatography-electrospray ionization-mass spectrometry (LC-ESI-MS/MS) method has been developed and validated for the identification and quantification of clebopride in human plasma using itopride as an internal standard. The method involves a simple liquid-liquid extraction. The analytes were separated by isocratic gradient elution on a CAPCELL MG-III C(18) (5 microm, 150 mm x 2.1 mm i.d.) column and analyzed in multiple reaction monitoring (MRM) mode with positive electrospray ionization (ESI) interface using the respective [M+H](+) ions, m/z 373.9-->m/z184.0 for clebopride, m/z 359.9-->m/z71.5 for itopride. The method was validated over the concentration range of 69.530-4450.0 pg/ml for clebopride. Within- and between-batch precision (RSD%) was all within 6.83% and accuracy ranged from -8.16 to 1.88%. The LLOQ was 69.530 pg/ml. The extraction recovery was on an average 77% for clebopride. The validated method was used to study the pharmacokinetics profile of clebopride in human plasma after oral administration of clebopride.