Merck
Lip plumper contact urticaria.
Journal of the American Academy of Dermatology (2009-04-25)
Elnaz F Firoz, Joshua M Levin, Rachael D Hartman, William D James
ABSTRACT
Lip plumpers are relatively new topical agents on the market that are designed to increase lip volume minutes to days after application. In this report, we describe a case of lip plumper contact urticaria in a young boy. We also discuss the mechanisms of action of lip plumpers and the sociocultural reasons for their use.
Sigma-Aldrich
Benzyl nicotinate, ≥98.0% (GC)