Merck
  • Alpha-fluoro-alpha-nitro(phenylsulfonyl)methane as a fluoromethyl pronucleophile: efficient stereoselective Michael addition to chalcones.
Alpha-fluoro-alpha-nitro(phenylsulfonyl)methane as a fluoromethyl pronucleophile: efficient stereoselective Michael addition to chalcones.
Proceedings of the National Academy of Sciences of the United States of America (2009-02-25)
G K Surya Prakash, Fang Wang, Timothy Stewart, Thomas Mathew, George A Olah
ABSTRACT
Highly efficient stereoselective 1,4-addition of racemic alpha-fluoro-alpha-nitro(phenylsulfonyl)methane (FNSM) as a fluoromethyl pronucleophile to alpha,beta-unsaturated ketones using a wide range of chiral organobifunctional catalysts under moderate conditions in the absence of an additional base has been achieved. A series of catalysts was screened for the enantioselective addition of FNSM to chalcones and the catalysts CN I, CD I, QN I-IV, and QD I were found to enable this reaction, successfully providing exclusive 1,4-addition products stereoselectively in high yields (conversion, diastereomeric ratio, and enantiomeric excess). Studies involving a model reaction and systematic analysis of the absolute configuration support the suggested mechanism.