MilliporeSigma
  • Redescription of the tintinnid Stenosemella pacifica Kofoid and Campbell, 1929 (Ciliophora, Spirotricha) based on live observation, protargol Impregnation, and Scanning Electron Microscopy.
Redescription of the tintinnid Stenosemella pacifica Kofoid and Campbell, 1929 (Ciliophora, Spirotricha) based on live observation, protargol Impregnation, and Scanning Electron Microscopy.
The Journal of eukaryotic microbiology (2008-03-06)
Sabine Agatha, Sheng-Fang Tsai
ABSTRACT
The tintinnid ciliate Stenosemella pacifica Kofoid and Campbell, 1929 was occasionally recorded from the pelagial of temperate, subtropical, and tropical neritic waters. Since its cytological features were unknown, the species is redescribed from material collected in the pelagial of the Irish Sea, using live observation, protargol impregnation, and scanning electron microscopy. Furthermore, the species diagnosis is improved to include new characteristics, e.g. the somatic ciliary pattern comprising a ventral, dorsal, and posterior kinety as well as a right, left, and lateral ciliary field. The stomatogenesis of S. pacifica is typical for species with such a complex somatic ciliary pattern: the oral primordium develops hypoapokinetally posterior to the lateral ciliary field. The presence of windows in the lorica collar of Stenosemella ventricosa, the type of the genus, necessitates (i) an improved genus diagnosis, (ii) a synonymization of the genus LuminellaKofoid and Campbell, 1939, and (iii) a transfer of the Luminella species to the genus Stenosemella, including Luminella neocalifornica, which becomes Stenosemella neocalifornica nov. comb. Owing to the lack of a description, Stenosemella crateri is considered a nomen nudum.
Sigma-Aldrich
Silver proteinate, ~8% Ag basis
Pricing and availability is not currently available.