Merck
  • Evaluation of proinflammatory cytokine pathway inhibitors for p38 MAPK inhibitory potential.
Evaluation of proinflammatory cytokine pathway inhibitors for p38 MAPK inhibitory potential.
Journal of medicinal chemistry (2007-11-09)
Rameshwar U Kadam, Divita Garg, Atish T Paul, K K Bhutani, Nilanjan Roy
ABSTRACT
The target for the anti-inflammatory natural products like amentoflavone ( 2), which act by interfering with the proinflammatory cytokine pathway (e.g., TNF-alpha, IL-1beta, and NO synthase), is not yet well-defined. Data obtained from docking, electronic, and surface analyses shed some light on steric and electronic complementarity of these molecules to p38 MAPK, thereby suggesting a possible mechanism by which they might reduce the production of proinflammatory cytokines.
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
Supelco
Luteolin 7-glucoside, analytical standard