Merck
  • Determination of the dimerization constant of pinacyanol: role of the thermochromic effect.
Determination of the dimerization constant of pinacyanol: role of the thermochromic effect.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-10-30)
Raimon Sabaté, Joan Estelrich
ABSTRACT
Pinacyanol (PIN), as other cyanine dyes, has demonstrated a unique ability to form associates such as dimers, and H- and J-aggregates. This association is strongly favoured in water, and even at low dye concentrations, dimers and superior order aggregates are present. As a consequence, the determination of the dimerization constant involves sometimes a significant error when these aggregates are neglected. As an increase in temperature shifts the equilibrium among the different species towards the lowest order aggregates, we have obtained the spectra of PIN at several temperatures. By extrapolating some spectral characteristics at high temperatures, a spectrum of the dimer without any contribution of other aggregates was obtained. From this spectrum and that of the monomer, the dimerization constant was calculated, as well as the Gibbs energy change associated to the reaction. The enthalpy and entropy changes of the dimerization were determined from the dependence of the dimerization constants on the temperature. From these results it can be inferred that the driving force of the dimerization is of enthalpic origin.
Sigma-Aldrich
1,1′-Diethyl-2,2′-carbocyanine iodide, 97%
Sigma-Aldrich
Pinacyanol chloride