Merck
  • Top-down solid-phase fabrication of nanoporous cadmium oxide architectures.
Top-down solid-phase fabrication of nanoporous cadmium oxide architectures.
Journal of the American Chemical Society (2007-02-06)
Haidong Yu, Deshen Wang, Ming-Yong Han
ABSTRACT
In this article, we have demonstrated one-step solid-phase transformation from high-quality cadmium carbonate microcrystals into highly nanoporous cadmium oxide. The high crystal quality of cadmium carbonate is critical for the successful fabrication of porous nanoarchitectures with predetermined morphology and well-controlled internal structure. This novel strategy has a good potential to prepare nanoporous materials at a large scale by using perfect monolithic carbonate crystals, and it is also useful to synthesize different nanoporous materials on metal-oxide-coated substrates. Meanwhile, this simple thermal transformation of cadmium carbonate into porous structures has further been extended to convert calcium carbonate into such porous structures.
Sigma-Aldrich
Cadmium carbonate, powder, 98%