- Synergistic regulation of cytosolic Ca2+ concentration in mouse astrocytes by NK1 tachykinin and adenosine agonists.
Synergistic regulation of cytosolic Ca2+ concentration in mouse astrocytes by NK1 tachykinin and adenosine agonists.
The effects on cytosolic Ca2+ concentration of 2-chloroadenosine and [L-Pro9]-substance P, a selective agonist of NK1 receptors, were investigated on astrocytes from embryonic mice in primary culture. Cells responded to [L-Pro9]-substance P with a transitory increase in cytosolic Ca2+ which was of shorter duration when external Ca2+ was removed. A transient response to 2-chloroadenosine alone occurred. When simultaneously applied, [L-Pro9]-substance P and 2-chloroadenosine evoked a prolonged elevation of cytosolic Ca2+ (up to 30 min). This phenomenon was dependent on the presence of extracellular Ca2+, but insensitive to dihydropyridines, La3+, and Co2+, excluding the implication of voltage-operated Ca2+ channels. Arachidonic acid also induced a sustained elevation of cytosolic Ca2+, but did not increase further the response evoked by [L-Pro9]-substance P and 2-chloroadenosine. The activation of protein kinase C by a diacylglycerol analogue mimicked the effect of [L-Pro9]-substance P in potentiating the 2-chloroadenosine-evoked response. Like 2-chloroadenosine, pinacidil, which hyperpolarizes the cells by opening K+ channels, prolonged the elevation of cytosolic Ca2+ concentration induced by [L-Pro9]-substance P. Conversely, depolarization with 50 mM KCl canceled the effects of either pinacidil or 2-chloroadenosine applied with [L-Pro9]-substance P. Pertussis toxin pretreatment suppressed all the effects induced by 2-chloroadenosine.