Merck
  • Protective effect of melatonin and pinoline on nitric oxide-induced lipid and protein peroxidation in rat brain homogenates.
Protective effect of melatonin and pinoline on nitric oxide-induced lipid and protein peroxidation in rat brain homogenates.
Neuroscience letters (2006-07-21)
G Piñol-Ripoll, L Fuentes-Broto, S Millán-Plano, M Reyes-Gonzáles, J A Mauri, E Martínez-Ballarín, R J Reiter, J J García
ABSTRACT
Nitric oxide (NO) is a physiological neurotransmitter, a mediator of the excitatory neurotransmitter glutamate pathways that regulates several neuroendocrine functions, but excessive NO is toxic by itself and it interacts with superoxide radical (O(2)(-)) to form the peroxynitrite anion (ONOO(-)). Using rat brain homogenates, we investigated the effects of melatonin and pinoline in preventing the level of lipid peroxidation (LPO) and carbonyl contents in proteins induced by nitric oxide (NO) which was released by the addition of sodium nitroprusside (SNP). Lipid and protein peroxidation were estimated by quantifying malondialdehyde (MDA) and 4-hydroxyalkenal (4-HDA) concentrations and carbonyl contents, respectively. SNP increased MDA+4-HDA and carbonyl contents production in brain homogenates in a time and concentration dependent manner. Both, melatonin and pinoline reduced NO-induced LPO and carbonyl contents in a dose-dependent manner in concentrations from 0.03 to 3 mM and 1 to 300 microM, respectively. Under the in vitro conditions of this experiment, both antioxidants were more efficient in limiting SNP protein oxidation than lipid damage.