Merck
  • Development of a gas chromatographic/mass spectrometric method to quantify R(-)-apomorphine, R(-)-apocodeine and R(-)-norapomorphine in human plasma and urine.
Development of a gas chromatographic/mass spectrometric method to quantify R(-)-apomorphine, R(-)-apocodeine and R(-)-norapomorphine in human plasma and urine.
Journal of mass spectrometry : JMS (2005-11-15)
Frédéric Libert, François Coudoré, Damien Richard, Franck Durif, Alain Eschalier
ABSTRACT
A method was developed and validated for the analysis of R(-)-apomorphine, (R-)-apocodeine and R(-)-norapomorphine in human plasma and urine with N-propylnorapomorphine as internal standard using gas chromatography/mass spectrometry (GC/MS) and single-ion monitoring after a single liquid-liquid extraction and silylation of compounds. The quantification limits were 1 ng/ml for apomorphine and apocodeine and 25 ng/ml for norapomorphine. Calibration curves were linear, within the range 1-100 ng/ml. Variation in intraday and interday precision was below 10%. This method was applied to study apomorphine bioavailability in nine patients with Parkinson's disease before and after coadministration of a catechol-O-methyl transferase inhibitor.