Merck
  • Fundamental adsorption characteristics of carbonaceous adsorbents for 1,2,3,4-tetrachlorobenzene in a model gas of an incineration plant.
Fundamental adsorption characteristics of carbonaceous adsorbents for 1,2,3,4-tetrachlorobenzene in a model gas of an incineration plant.
Environmental science & technology (2005-08-30)
Kenichiro Inoue, Katsuya Kawamoto
ABSTRACT
Carbonaceous adsorbents such as activated carbon have been used to reduce the emission of organic pollutants from incineration plants. However, with this method, the amount and type of adsorbent to be used are based only on empirical results, which may lead to overuse of the adsorbents. The fundamental adsorption characteristics of several kinds of activated carbon, activated coke, and carbide wood were examined using 1 ,2,3,4-tetrachlorobenzene as an adsorbate. The removal performance and various equilibrium adsorption characteristics of these adsorbents were analyzed using laboratory-scale adsorption equipment. The equilibrium adsorption amount increased by a factor of 1.9-3.2 at 150 degrees C compared with that at 190 degrees C. The effect of the moisture content on adsorption capacity was relatively small in comparison with that of the temperature. The micropore volume for pore diameters of 2 nm or less was the most important factor governing the adsorption capacity for all adsorbents. Activated carbon showed superior adsorption ability compared to activated coke and carbide wood, although all adsorbents were sufficient for practical use.
Sigma-Aldrich
1,2,3,4-Tetrachlorobenzene, 98%