Merck
Encapsulated liquid crystals as probes for remote thermometry.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group (1992-03-01)
K J Franklin, R J Buist, J den Hartog, G A McRae, D P Spencer
ABSTRACT
A temperature probe based on the magnetic resonance properties of an encapsulated liquid crystal has been investigated. Large changes in magnetic resonance signals occur as the liquid crystal undergoes a phase transition from an anisotropic (nematic) state to the isotropic liquid. The low latent heat of such phase transitions allows for rapid phase changes during a hyperthermia treatment. Transition temperatures can be tailored by adding suitable compounds such as analogues of the liquid crystal or various solvents. Encapsulation is required to maintain the integrity of the liquid crystal, particularly for applications in vivo. Results of preliminary studies designed to demonstrate the technical feasibility of the concept are presented.