Merck
  • Hydromorphone metabolites: isolation and identification from pooled urine samples of a cancer patient.
Hydromorphone metabolites: isolation and identification from pooled urine samples of a cancer patient.
Xenobiotica; the fate of foreign compounds in biological systems (2002-06-18)
M Zheng, K M McErlane, M C Ong
ABSTRACT
1. Hydromorphone-3-glucuronide, dihydromorphine, dihydroisomorphine, dihydromorphine-3-glucuronide and dihydroisomorphine-3-glucuronide were isolated from a cancer patient's urine and identified as metabolites of hydromorphone by comparison with synthetic standards using LC/MS/MS with gradient elution. 2. The relative urinary recovery of dihydroisomorphine-3-glucuronide was estimated to be 17-fold higher than previously reported. 3. Three new metabolites, including hydromorphone-3-sulphate, norhydromorphone and nordihydroisomorphine, were tentatively identified.
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclohexanone, 99.8%
Supelco
Cyclohexanone, Selectophore, ≥99.5%