Merck
  • Immunization with gp96 from Listeria monocytogenes-infected mice is due to N-formylated listerial peptides.
Immunization with gp96 from Listeria monocytogenes-infected mice is due to N-formylated listerial peptides.
Journal of immunology (Baltimore, Md. : 1950) (2001-11-21)
A M Sponaas, U Zuegel, S Weber, R Hurwitz, R Winter, S Lamer, P R Jungblut, S H Kaufmann
ABSTRACT
N-Formylated (N-f-met) peptides derived from proteins of the intracellular bacterium Listeria monocytogenes generate a protective, H2-M3-restricted CD8 T cell response in C57BL/6 mice. N-f-met peptide-specific CTL were generated in vitro when mice previously immunized with gp96 isolated from donor mice infected with L. monocytogenes were stimulated with these peptides. No significant peptide-specific CTL activity was observed in mice immunized with gp96 from uninfected animals. Masses corresponding to one N-f-met peptide were found by matrix-assisted laser desorption/ionization-mass spectrometry on gp96 isolated from C57BL/6 mice infected with L. monocytogenes, but not on gp96 from noninfected mice. Therefore, bacterial N-f-met peptides from intracellular bacteria can bind to gp96 in the infected host, and gp96 loaded with these peptides can generate N-f-met-peptide-specific CTL. We assume a unique role of gp96 in Ag processing through the H2-M3 pathway.
Sigma-Aldrich
N-Formyl-L-methionine, ≥90% (TLC)