Merck
In vitro selection of DNA aptamers that bind L-tyrosinamide.
Bioorganic & medicinal chemistry (2001-09-15)
E Vianini, M Palumbo, B Gatto
ABSTRACT
We have applied SELEX (Systematic Evolution of Ligands by EXponential enrichment), a combinatorial method that employs biopolymers for drug discovery, to identify single stranded DNA sequences able to bind L-Tyrosinamide, a simple mimic of Tyrosine, an amino acid essential to the catalytic activity of several enzymes of pharmaceutical interest. After 15 SELEX cycles using L-Tyrosinamide immobilized on an affinity chromatography column, the percentage of aptamers specifically eluted from the affinity column with free L-Tyrosinamide was 55% of the total. Aptamers were subcloned and sequenced, allowing the identification of a highly conserved consensus sequence, and showed a K(d) value for L-Tyrosinamide of 45 microM. The identified aptamer sequence will constitute the basis for further in vitro evolution protocols and structure-based drug design.