Merck
  • Novel C-terminus modifications of the Dmt-Tic motif: a new class of dipeptide analogues showing altered pharmacological profiles toward the opioid receptors.
Novel C-terminus modifications of the Dmt-Tic motif: a new class of dipeptide analogues showing altered pharmacological profiles toward the opioid receptors.
Journal of medicinal chemistry (2001-07-13)
D Pagé, A Naismith, R Schmidt, M Coupal, M Labarre, M Gosselin, D Bellemare, K Payza, W Brown
ABSTRACT
The design, synthesis and pharmacological evaluation of a novel class of Dmt-Tic dipeptide analogues are described. These resulting analogues bearing different C-terminal functionalities were found to bind to the human delta receptor with high affinity. One specific class of dipeptides bearing urea/thiourea functionalities showed partial to full activation of the delta receptor. Several dipeptides also showed good binding affinities with full activation of the human kappa receptor, a novel property for those ligands.
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%