Merck
  • Optimization of N-benzyl-benzoxazol-2-ones as receptor antagonists of macrophage migration inhibitory factor (MIF).
Optimization of N-benzyl-benzoxazol-2-ones as receptor antagonists of macrophage migration inhibitory factor (MIF).
Bioorganic & medicinal chemistry letters (2010-08-24)
Alissa A Hare, Lin Leng, Sunilkumar Gandavadi, Xin Du, Zoe Cournia, Richard Bucala, William L Jorgensen
ABSTRACT
The cytokine MIF is involved in inflammation and cell proliferation via pathways initiated by its binding to the transmembrane receptor CD74. MIF also exhibits keto-enol tautomerase activity, believed to be vestigial in mammals. Starting from a 1 μM hit from virtual screening, substituted benzoxazol-2-ones have been discovered as antagonists with IC(50) values as low as 7.5 nM in a tautomerase assay and 80 nM in a MIF-CD74 binding assay. Additional studies for one of the potent inhibitors demonstrated that it is not a covalent inhibitor of MIF and that it attenuates MIF-dependent ERK1/2 phosphorylation in human synovial fibroblasts.