Merck
  • Suppression of the blowfly Lucilia sericata using odour-baited triflumuron-impregnated targets.
Suppression of the blowfly Lucilia sericata using odour-baited triflumuron-impregnated targets.
Medical and veterinary entomology (1998-11-24)
K E Smith, R Wall
ABSTRACT
Field trials were carried out in 1995 and 1996 on farms in the south-west of England to assess the extent to which odour-baited targets could be used to suppress populations of the ectoparasitic blowfly, Lucilia sericata, in sheep pastures. Targets were constructed from 41 x 41 cm squares of aluminium sheet, covered by white cloth which had been dipped in a mixture of sucrose solution (50% w/v) and the chitin synthesis inhibitor triflumuron (10% suspension concentrate). Each target was baited with approximately 300 g of liver and sodium sulphide solution (10%). Three matched sheep farms were used in the trials. In 1995, triflumuron-impregnated targets were placed around the periphery of sheep pastures at one of the farms in late June, at approximately one target per hectare. In 1996, triflumuron-impregnated targets were placed around the periphery of sheep pastures of a second of the farms in early May, at approximately five targets per hectare. Each year, five sticky targets, used to monitor the L. sericata populations, were also placed in fields at the experimental and the other two farms, which acted as controls. In 1995, the results provided some, although inconclusive, evidence that the triflumuron-impregnated targets had reduced the numbers of L. sericata relative to the populations on the two control farms. In 1996, however, the density of L. sericata on the experimental farm was reduced to almost zero and remained significantly lower than on two control farms throughout the period during which the triflumuron-impregnated targets remained in the field. The results are discussed in relation to the use of triflumuron-treated targets as a practical means of controlling L. sericata and sheep blowfly strike.
Product Description
Supelco
Triflumuron, PESTANAL®, analytical standard