Merck
  • [Palladium-containing compounds possess radiation-protective properties and enhance adaptive response of CFUs to radiation].
[Palladium-containing compounds possess radiation-protective properties and enhance adaptive response of CFUs to radiation].
Radiatsionnaia biologiia, radioecologiia (1998-02-19)
T N Semenets, O V Semina, I A Efimenko, N A Ivanova, A M Poverennyĭ
ABSTRACT
Efasol and potassium tetrachloropalladate being injected into mice prior to irradiation with a dose of 150 sGy increase the colony forming activity of animals bone marrow cells transplanted in lethally irradiated recipient mice. The palladium presence in such compounds is supposed to be a reason of radioprotection. Upon the combined application of potassium tetrachloropalladate and irradiation of mice with an adaptive dose of 5 sGy a potentiation of individual effect of each separate agent to increase CFUs resistance against the consequent irradiation was observed.
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis