Merck
  • Esterification of 9-fluorenylmethoxycarbonyl-glycosylated serine and cysteine derivatives with an hydroxymethyl resin.
Esterification of 9-fluorenylmethoxycarbonyl-glycosylated serine and cysteine derivatives with an hydroxymethyl resin.
The journal of peptide research : official journal of the American Peptide Society (1998-01-24)
E Harth-Fritschy, D Cantacuzène
ABSTRACT
Esterification of glycosylated serine and cysteine derivatives with a 4-alkoxybenzyl alcohol (Wang) resin is described. The classical methods of ester bond formation (symmetrical anhydride, 2-(1H-benzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate [TBTU]/4-dimethylaminopyridine [DMAP] with or without 1-hydroxybenzotriazole [HOBT], pentafluorophenyl [Pfp] esters gave high percentages of racemization of the glycosylated serine or cysteine residues. To reduce the D-amino acid content, we found that the best results were obtained with the highly efficient MSNT reagent (2,4,6-mesitylenesulfonyl-3-nitro-1,2,4-triazolide), which gave a high yield of substitution of the resin and the lowest percentage of racemization. A difference in behavior was observed between the two amino acids. The glycosylated cysteine derivative always gave lower racemization than the analogous glycosylated serine.
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Pricing and availability is not currently available.
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, for chiral derivatization, LiChropur, ≥99.0%
Pricing and availability is not currently available.