Merck
  • Occupational dermatoses from exposure to epoxy resin compounds in a ski factory.
Occupational dermatoses from exposure to epoxy resin compounds in a ski factory.
Contact dermatitis (1996-06-01)
R Jolanki, K Tarvainen, T Tatar, T Estlander, M L Henriks-Eckerman, K K Mustakallio, L Kanerva
ABSTRACT
Of 22 workers in a ski factory, occupational allergic contact dermatitis was found in 8. 6 were sensitive to epoxy resin compounds, i.e., epoxy resins, hardeners or diluents, 1 to cobalt in glass-fiber reinforcements, and 1 to formaldehyde in a urea-formaldehyde glue and a lacquer. 4 workers had irritant contact dermatitis from epoxy resin compounds, lacquers, sanding dust, or glass-fiber dust. 3 had contact allergy from a new sensitizer, diethyleneglycol diglycidyl ether, in a reactive diluent. Immediate transfer of workers sensitized to epoxy resin from epoxy exposure prevents aggravation of their dermatitis and broadening of the sensitization to epoxy hardeners, diluents and other compounds.
Product Description
Supelco
Diethylene glycol dimethyl ether, analytical standard
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%