Merck
  • Sensitive high-performance liquid chromatographic determination of cyclizine and its demethylated metabolite, norcyclizine, in biological fluids using coulometric detection.
Sensitive high-performance liquid chromatographic determination of cyclizine and its demethylated metabolite, norcyclizine, in biological fluids using coulometric detection.
Journal of chromatography. B, Biomedical applications (1995-10-06)
R B Walker, I Kanfer
ABSTRACT
An accurate, sensitive, selective and reproducible high-performance liquid chromatographic method with coulometric detection for the determination of cyclizine and its inactive demethylated metabolite, norcyclizine, in biological fluids has been developed. The drugs were separated using a custom packed reversed-phase C18 analytical column and phosphate buffer (0.05 M, pH 3)-acetonitrile (7:3) as mobile phase. The dual electrode coulometric detector was operated in the "oxidative-screen" mode with the upstream electrode (detector 1) set at 0.55 V and the downstream electrode (detector 2) set at 0.90 V. Serum and urine samples were prepared for analysis by solid-phase extraction, followed by a simple phase-separation step. The limit of quantitation was 1 ng/ml for both cyclizine and norcyclizine in serum and urine.