Merck
  • Preparation, characterization, and pharmaceutical application of linear dextrins. II. Complexation and dispersion of drugs with amylodextrin by freeze-drying and kneading.
Preparation, characterization, and pharmaceutical application of linear dextrins. II. Complexation and dispersion of drugs with amylodextrin by freeze-drying and kneading.
Pharmaceutical research (1993-09-01)
G H Te Wierik, A C Eissens, A C Besemer, C F Lerk
ABSTRACT
The ability of amylodextrin (a linear dextrin) to act as a complexing agent or as a carrier for solid dispersion was evaluated. Blends of amylodextrin with diazepam or prednisolone were freeze-dried and kneaded at elevated temperatures, respectively. The products were analyzed by DSC, X-ray diffractometry, and FTIR spectroscopy. Complex formation with amylodextrin by freeze-drying was found not to occur for diazepam but for prednisolone at a molar ratio of 1 to 1. The freeze-dried product of diazepam with amylodextrin proved to be a solid dispersion. Solid dispersions were formed by both wet (with ethanol) and dry kneading at elevated temperatures of low-melting drugs such as lidocain, diazepam, and methyl-PABA with amylodextrin. No solid dispersions were obtained for high-melting drugs such as prednisolone and salicylic acid. The results point to the formation of solid dispersions by a melting mechanism during the process of kneading at elevated temperatures of low-melting drugs with amylodextrin.
Sigma-Aldrich
Starch, soluble, ACS reagent