Merck
  • Inherited 3-methylglutaconic aciduria in two brothers--another defect of leucine metabolism.
Inherited 3-methylglutaconic aciduria in two brothers--another defect of leucine metabolism.
The Journal of pediatrics (1982-10-01)
M Duran, F A Beemer, A S Tibosch, L Bruinvis, D Ketting, S K Wadman
ABSTRACT
Two brothers, aged 7 and 5 years, who excreted large amounts of the leucine metabolites 3-methylglutaconic acid, 3-methylglutaric acid, and 3-hydroxyisovaleric acid, are described. The excretion of these metabolites could be enhanced by increasing the leucine intake. Restriction of the protein intake resulted in a marked reduction of the metabolite excretion. However, the excretion of the ultimate leucine metabolite, 3-hydroxy-3-methylglutaric acid, remained unchanged at a low level. The only clinical abnormality was speech retardation. A (partial) deficiency of 3-methylglutaconyl coenzyme A hydratase is proposed to be the most likely underlying defect.
Sigma-Aldrich
3-Methylglutaric acid, 99%