- Carboxypeptidase activity in human mycoplasmas.
Carboxypeptidase activity in human mycoplasmas.
Journal of bacteriology (1986-11-01)
K Shibata, T Watanabe
PMID3096956
ABSTRACT
Mycoplasma salivarium produced citrulline, ammonia, and ATP from N-benzoylglycyl-L-arginine. The activity was inhibited by EDTA and was therefore concluded to be due to an arginine-specific carboxypeptidase. The activity was also found to exist in M. orale, M. buccale, M. faucium, and M. hominis.
MATERIALS