Merck
  • ZnO nanoparticles induces cell death in malignant human T98G gliomas, KB and non-malignant HEK cells.
ZnO nanoparticles induces cell death in malignant human T98G gliomas, KB and non-malignant HEK cells.
Journal of biomedical nanotechnology (2013-08-06)
Rizwan Wahab, Nagendra Kumar Kaushik, Neha Kaushik, Eun Ha Choi, Ahmad Umar, Sourabh Dwivedi, Javed Musarrat, Abdulaziz A Al-Khedhairy
ABSTRACT
This paper reports the synthesis and characterization of ZnO nanoparticles prepared by soft chemical process. The nanoparticles of ZnO possess wurtzite hexagonal phase and were used for the induction of cell death in malignant human T98G gliomas, KB epithermoids and HEK normal non-malignant kidney cells. By applying ZnO nanoparticles, the cells exhibit that the nanoparticles are more efficacious on T98G cancer cells, moderately effective on KB cells and least toxic on normal human HEK cells. The results demonstrated that the treatment with ZnO nanoparticles sensitize T98G cells by increasing both the mitotic (linked to cytogenetic damage) and interphase (apoptosis) death. The ZnO nanoparticles behave as genotoxic drugs, since they induce a micronucleus formation in cells. The present study could be helpful in designing more potent anticancer agents for the therapeutic uses.
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%