Merck
  • Fabrication of seamless electrospun collagen/PLGA conduits whose walls comprise highly longitudinal aligned nanofibers for nerve regeneration.
Fabrication of seamless electrospun collagen/PLGA conduits whose walls comprise highly longitudinal aligned nanofibers for nerve regeneration.
Journal of biomedical nanotechnology (2013-07-19)
Yuanming Ouyang, Chen Huang, Yi Zhu, Cunyi Fan, Qinfei Ke
ABSTRACT
An ideal nerve scaffold should supply structural guidance and trophic support to facilitate nerve regeneration. Aligned electrospun nanofibers have shown considerable promise for the precise guidance of regenerating axons in vitro and in vivo. Therefore, uniaxially aligned three-dimension (3D) nanofiberous scaffolds may allow regenerating axons to traverse large gaps to treat severe nerve injuries. However, the aligned 3D conduit was always rolled by an aligned 2-dimensional (2D) sheet in current fabrication methods, which was inconvenient for transplant due to the discontinuous joint and inconsistent size. We developed a modified one-step electrospinning technique to produce a seamless 3D nanofiberous nerve conduit (NC) with highly longitudinal aligned nanofibers that combines the biocompatibility of natural collagen and the strength of the synthetic polymer poly(lactic-co-glycolic acid) (PLGA). Scanning electron microscopy (SEM) confirmed the parallel alignment of the scaffold fibers. To test the effectiveness of these scaffolds at restoring neuronal connections, they were implanted into adult rats across a 13 mm sciatic nerve defect. Tests of, motor function, nerve conduction, axonal and Schwann cell morphology, and marker expression all revealed that uniaxially aligned seamless 3D electrospun collagen/PLGA NCs were superior to randomly oriented NCs and inferior to autografts for promoting axon regeneration, myelination, action potential propagation, neuromuscular transmission, and functional recovery. These uniaxially aligned seamless 3D electrospun collagen/PLGA nerve guides can also incorporate signaling molecules and additional structural cues to guide nerve growth, and so may be a promising substitute for autogenous nerve grafts.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance